missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminophenylacetic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.
Solubility
Very soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.Stable under normal temperatures and pressures. Stable at room temperature in closed containers under normal storage and handling conditions.
4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.
Solubility
Very soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.Stable under normal temperatures and pressures. Stable at room temperature in closed containers under normal storage and handling conditions.
Specifications
Melting Point | ∼201°C (decomposition) |
Beilstein | 2208094 |
Merck Index | 14,463 |
Solubility Information | Very soluble in water. |
Formula Weight | 151.17 |
Percent Purity | 98% |
Quantity | 50 g |
Chemical Name or Material | 4-Aminophenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'