missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminophenylacetic acid, 98%, ACROS Organics™
£15.37 - £163.00
Chemical Identifiers
CAS | 1197-55-3 |
---|---|
Molecular Formula | C8H9NO2 |
Molecular Weight (g/mol) | 151.16 |
MDL Number | MFCD00007916 |
InChI Key | CSEWAUGPAQPMDC-UHFFFAOYSA-N |
Synonym | 4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid |
PubChem CID | 14533 |
IUPAC Name | 2-(4-aminophenyl)acetic acid |
SMILES | C1=CC(=CC=C1CC(=O)O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10018661
|
Acros Organics
104280050 |
5g | Glass bottle |
£15.37
5g |
Estimated Shipment 03-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10020630
|
Acros Organics
104280250 |
25g | Glass bottle |
£46.11
25g |
Estimated Shipment 04-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10177503
|
Acros Organics
104281000 |
100g | Glass bottle |
£163.00
100g |
In stock, ready to ship: 1 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
1197-55-3 | |
151.16 | |
CSEWAUGPAQPMDC-UHFFFAOYSA-N | |
14533 | |
C1=CC(=CC=C1CC(=O)O)N |
C8H9NO2 | |
MFCD00007916 | |
4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid | |
2-(4-aminophenyl)acetic acid |
Specifications
4-Aminophenylacetic acid | |
97.5 | |
97.5% min. (HPLC) | |
H2NC6H4CH2CO2H | |
MFCD00007916 | |
15, 459 | |
Glass bottle | |
CSEWAUGPAQPMDC-UHFFFAOYSA-N | |
2-(4-aminophenyl)acetic acid | |
14533 | |
Powder | |
1% max. (K.F.) | |
196.0°C to 201.0°C |
1197-55-3 | |
100.0 | |
C8H9NO2 | |
Authentic | |
14, 456 | |
4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid | |
Solubility in water: insoluble. Other solubilities: soluble in alkalies, soluble in alcohol | |
C1=CC(=CC=C1CC(=O)O)N | |
151.16 | |
151.16 | |
98% | |
Yellow to Beige | |
5g |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Aminophenylacetic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.