missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenylacetic acid, 98%, Thermo Scientific™

£17.80 - £238.00

Chemical Identifiers

CAS 1197-55-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007916
InChI Key CSEWAUGPAQPMDC-UHFFFAOYSA-N
Synonym 4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid
PubChem CID 14533
IUPAC Name 2-(4-aminophenyl)acetic acid
SMILES NC1=CC=C(CC(O)=O)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11437656
SDS View Documents
Thermo Scientific Alfa Aesar
A14675.09
10g
£17.80
10g
View alternative products
Save Up to 
»
In stock, ready to ship: 1
Please to purchase this item. Need a web account? Register with us today!
11447656
SDS View Documents
Thermo Scientific Alfa Aesar
A14675.18
50g
£57.00
50g
View alternative products
Save Up to 
»
In stock, ready to ship: 4
Please to purchase this item. Need a web account? Register with us today!
11457656
SDS View Documents
Thermo Scientific Alfa Aesar
A14675.30
250g
£238.00
250g
View alternative products
Save Up to 
»
Estimated Shipment: 04-11-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications
4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.Stable under normal temperatures and pressures. Stable at room temperature in closed containers under normal storage and handling conditions.
Specifications

Chemical Identifiers

1197-55-3
151.17
CSEWAUGPAQPMDC-UHFFFAOYSA-N
14533
NC1=CC=C(CC(O)=O)C=C1
C8H9NO2
MFCD00007916
4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid
2-(4-aminophenyl)acetic acid

Specifications

4-Aminophenylacetic acid
C8H9NO2
2208094
4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid
CSEWAUGPAQPMDC-UHFFFAOYSA-N
2-(4-aminophenyl)acetic acid
14533
98%
10g
1197-55-3
MFCD00007916
14,463
Very soluble in water.
NC1=CC=C(CC(O)=O)C=C1
151.17
151.17
∼201°C (decomposition)
SDS
Product Certifications

RUO – Research Use Only