missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenyl ether, 98%, Thermo Scientific Chemicals

Catalog Number 10459200
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
101-80-4
C12H12N2O
200.24
MFCD00007863
HLBLWEWZXPIGSM-UHFFFAOYSA-N
4,4'-oxydianiline, 4,4'-diaminodiphenyl ether, 4,4-oxydianiline, 4-aminophenyl ether, 4,4'-oxybisbenzenamine, oxydianiline, 4-4-aminophenoxy aniline, diaminodiphenyl ether, benzenamine, 4,4'-oxybis, p,p'-oxydianiline
7579
CHEBI:34384
4-(4-aminophenoxy)aniline
C1=CC(=CC=C1N)OC2=CC=C(C=C2)N
This item is not returnable. View return policy
101-80-4
C12H12N2O
200.24
MFCD00007863
HLBLWEWZXPIGSM-UHFFFAOYSA-N
4,4'-oxydianiline, 4,4'-diaminodiphenyl ether, 4,4-oxydianiline, 4-aminophenyl ether, 4,4'-oxybisbenzenamine, oxydianiline, 4-4-aminophenoxy aniline, diaminodiphenyl ether, benzenamine, 4,4'-oxybis, p,p'-oxydianiline
7579
CHEBI:34384
4-(4-aminophenoxy)aniline
C1=CC(=CC=C1N)OC2=CC=C(C=C2)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 189°C to 193°C
Boiling Point 190°C (0.1 mmHg)
Flash Point 218°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (H2NC6H4)2O
Quantity 5 g
Beilstein 13, 441
Solubility Information Solubility in water: negligible
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material 4-Aminophenyl ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Aminophenyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'