missing translation for 'onlineSavingsMsg'
Learn More

4-Aminonicotinic acid, 97%, Thermo Scientific™

£21.00 - £64.10

Chemical Identifiers

CAS 7418-65-7
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00234183
InChI Key IASBMUIXBJNMDW-UHFFFAOYSA-N
Synonym 4-aminonicotinic acid, 4-amino-3-pyridinecarboxylic acid, 4-amino-nicotinic acid, 4-amino-3-carboxypyridine, 4-aminonicotinicacid, 4-amino nicotinic acid, 3-pyridinecarboxylic acid, 4-amino, 4-amino-3-pyridine carboxylic acid, zlchem 370, pubchem1214
PubChem CID 319979
ChEBI CHEBI:68577
IUPAC Name 4-aminopyridine-3-carboxylic acid
SMILES C1=CN=CC(=C1N)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10702453
SDS View Documents
Thermo Scientific Acros
361130010
1g Glass bottle
£21.00
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 8
Please to purchase this item. Need a web account? Register with us today!
10083383
SDS View Documents
Thermo Scientific Acros
361130050
5g Glass bottle
£64.10
5g
View alternative products
Save Up to 
»
Estimated Shipment: 14-12-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7418-65-7
138.13
IASBMUIXBJNMDW-UHFFFAOYSA-N
319979
4-aminopyridine-3-carboxylic acid
C6H6N2O2
MFCD00234183
4-aminonicotinic acid, 4-amino-3-pyridinecarboxylic acid, 4-amino-nicotinic acid, 4-amino-3-carboxypyridine, 4-aminonicotinicacid, 4-amino nicotinic acid, 3-pyridinecarboxylic acid, 4-amino, 4-amino-3-pyridine carboxylic acid, zlchem 370, pubchem1214
CHEBI:68577
C1=CN=CC(=C1N)C(=O)O

Specifications

4-Aminonicotinic acid
Glass bottle
1g
96.0
97%
MFCD00234183
IASBMUIXBJNMDW-UHFFFAOYSA-N
4-aminopyridine-3-carboxylic acid
319979
138.13
97%
Authentic
Cream to Yellow
7418-65-7
100.0
C6H6N2O2
4-aminonicotinic acid, 4-amino-3-pyridinecarboxylic acid, 4-amino-nicotinic acid, 4-amino-3-carboxypyridine, 4-aminonicotinicacid, 4-amino nicotinic acid, 3-pyridinecarboxylic acid, 4-amino, 4-amino-3-pyridine carboxylic acid, zlchem 370, pubchem1214
C1=CN=CC(=C1N)C(=O)O
138.13
CHEBI:68577
Powder
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Aminonicotinic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Product Certifications

RUO – Research Use Only