missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminonaphthalene-1-sulfonic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Amino-1-naphthalenesulfonic acid has potent therapeutic applications. Its sodium salt has been used as non-toxic hemostatic. It is also a metabolite of azo dyes.
Solubility
Sparingly soluble in dichloromethane. Soluble in aqueous base.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
4-Amino-1-naphthalenesulfonic acid has potent therapeutic applications. Its sodium salt has been used as non-toxic hemostatic. It is also a metabolite of azo dyes.
Solubility
Sparingly soluble in dichloromethane. Soluble in aqueous base.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | >300°C |
Quantity | 25 g |
UN Number | UN2585 |
Beilstein | 1971299 |
Merck Index | 14,6403 |
Solubility Information | Sparingly soluble in dichloromethane. Soluble in aqueous base. |
Formula Weight | 223.25 |
Percent Purity | 98% |
Chemical Name or Material | 4-Aminonaphthalene-1-sulfonic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'