missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethyltetrahydropyran, 97%, Thermo Scientific Chemicals

Catalog Number p-3586364
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
This item is not returnable. View return policy
130290-79-8
C6H13NO
115.17
MFCD02179435
IPBPLHNLRKRLPJ-UHFFFAOYSA-N
4-aminomethyltetrahydropyran, 4-aminomethyl tetrahydro-2h-pyran, tetrahydro-2h-pyran-4-yl methanamine, 4-aminomethyl tetrahydropyran, 1-tetrahydro-2h-pyran-4-ylmethanamine, tetrahydropyran-4-yl methylamine, c-tetrahydro-pyran-4-yl-methylamine, tetrahydro-2h-pyran-4-yl methylamine, 2h-pyran-4-methanamine, tetrahydro, oxan-4-yl methanamine
2773210
oxan-4-ylmethanamine
C1COCCC1CN
This item is not returnable. View return policy
130290-79-8
C6H13NO
115.17
MFCD02179435
IPBPLHNLRKRLPJ-UHFFFAOYSA-N
4-aminomethyltetrahydropyran, 4-aminomethyl tetrahydro-2h-pyran, tetrahydro-2h-pyran-4-yl methanamine, 4-aminomethyl tetrahydropyran, 1-tetrahydro-2h-pyran-4-ylmethanamine, tetrahydropyran-4-yl methylamine, c-tetrahydro-pyran-4-yl-methylamine, tetrahydro-2h-pyran-4-yl methylamine, 2h-pyran-4-methanamine, tetrahydro, oxan-4-yl methanamine
2773210
oxan-4-ylmethanamine
C1COCCC1CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.0200g/mL
Boiling Point 75.0°C to 76.0°C (18.0 mmHg)
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4640 to 1.4660
Quantity 250 mg
Specific Gravity 1.02
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material 4-Aminomethyltetrahydropyran
compliance-icons
Product Identifier
  • 1-tetrahydro-2H-pyran-4-ylmethanamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'