missing translation for 'onlineSavingsMsg'
Learn More

4-Aminocarbonylphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number 11926091
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
123088-59-5
C7H8BNO3
164.96
MFCD03411940
GNRHNKBJNUVWFZ-UHFFFAOYSA-N
4-aminocarbonylphenylboronic acid, 4-carbamoylphenyl boronic acid, benzamide-4-boronic acid, 4-aminocarbonyl benzeneboronic acid, 4-carbamoylbenzeneboronic acid, 4-aminocarbonylphenyl boronic acid, 4-dihydroxyboranyl benzamide, p-aminocarbonyl phenylboronic acid, boronic acid, 4-aminocarbonyl phenyl, 4-aminocarbonyl phenylboronic acid
2737811
(4-carbamoylphenyl)boronic acid
NC(=O)C1=CC=C(C=C1)B(O)O
This item is not returnable. View return policy
123088-59-5
C7H8BNO3
164.96
MFCD03411940
GNRHNKBJNUVWFZ-UHFFFAOYSA-N
4-aminocarbonylphenylboronic acid, 4-carbamoylphenyl boronic acid, benzamide-4-boronic acid, 4-aminocarbonyl benzeneboronic acid, 4-carbamoylbenzeneboronic acid, 4-aminocarbonylphenyl boronic acid, 4-dihydroxyboranyl benzamide, p-aminocarbonyl phenylboronic acid, boronic acid, 4-aminocarbonyl phenyl, 4-aminocarbonyl phenylboronic acid
2737811
(4-carbamoylphenyl)boronic acid
NC(=O)C1=CC=C(C=C1)B(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 242.0°C to 252.0°C
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 164.96
Percent Purity 97%
Chemical Name or Material 4-Aminocarbonylphenylboronic acid
compliance-icons
Product Identifier
  • 4-Aminocarbonylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'