missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzotrifluoride, 99%, Thermo Scientific Chemicals

Catalog Number p-55935
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
455-14-1
C7H6F3N
161.13
MFCD00064396
ODGIMMLDVSWADK-UHFFFAOYSA-N
4-trifluoromethyl aniline, 4-aminobenzotrifluoride, p-aminobenzotrifluoride, p-trifluoromethylaniline, benzenamine, 4-trifluoromethyl, 4-trifluoromethyl benzenamine, p-trifluoromethyl aniline, aniline, p-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluidine, a,a,a-trifluoro-p-toluidine
9964
CHEBI:40750
4-(trifluoromethyl)aniline
C1=CC(=CC=C1C(F)(F)F)N
This item is not returnable. View return policy
455-14-1
C7H6F3N
161.13
MFCD00064396
ODGIMMLDVSWADK-UHFFFAOYSA-N
4-trifluoromethyl aniline, 4-aminobenzotrifluoride, p-aminobenzotrifluoride, p-trifluoromethylaniline, benzenamine, 4-trifluoromethyl, 4-trifluoromethyl benzenamine, p-trifluoromethyl aniline, aniline, p-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluidine, a,a,a-trifluoro-p-toluidine
9964
CHEBI:40750
4-(trifluoromethyl)aniline
C1=CC(=CC=C1C(F)(F)F)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 5.0°C to 8.0°C
Color Colorless to Yellow
Density 1.2960g/mL
Boiling Point 83.0°C (12.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CF3
Refractive Index 1.4830 to 1.4850
Quantity 10 mL
Beilstein 12, III, 2151
Specific Gravity 1.296
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ether,methanol,ethanol and,aromatics
Formula Weight 161.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Aminobenzotrifluoride, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Aminobenzotrifluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'