missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzonitrile, 98%, ACROS Organics™

£21.40 - £236.00

Chemical Identifiers

CAS 873-74-5
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00007821
InChI Key YBAZINRZQSAIAY-UHFFFAOYSA-N
Synonym 4-cyanoaniline, p-aminobenzonitrile, p-cyanoaniline, benzonitrile, 4-amino, aniline, p-cyano, benzonitrile, p-amino, 1-amino-4-cyanobenzene, unii-pb8v69y76t, 4-amino benzonitrile, 4-amino-benzonitrile
PubChem CID 13396
IUPAC Name 4-aminobenzonitrile
SMILES C1=CC(=CC=C1C#N)N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10206220
SDS View Documents
Acros Organics
164470100
10g Glass bottle
£21.40
10g
View alternative products
Save Up to 
»
Estimated Shipment: 11-10-2021
Please to purchase this item. Need a web account? Register with us today!
10337680
SDS View Documents
Acros Organics
164470500
50g Glass bottle
£67.00
50g
View alternative products
Save Up to 
»
Estimated Shipment: 11-10-2021
Please to purchase this item. Need a web account? Register with us today!
10023423
SDS View Documents
Acros Organics
164472500
250g Glass bottle
£236.00
250g
View alternative products
Save Up to 
»
Estimated Shipment: 11-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

873-74-5
118.14
YBAZINRZQSAIAY-UHFFFAOYSA-N
13396
C1=CC(=CC=C1C#N)N
C7H6N2
MFCD00007821
4-cyanoaniline, p-aminobenzonitrile, p-cyanoaniline, benzonitrile, 4-amino, aniline, p-cyano, benzonitrile, p-amino, 1-amino-4-cyanobenzene, unii-pb8v69y76t, 4-amino benzonitrile, 4-amino-benzonitrile
4-aminobenzonitrile

Specifications

4-Aminobenzonitrile
Glass bottle
83.0°C to 87.0°C
873-74-5
100.0
C7H6N2
MFCD00007821
4-cyanoaniline, p-aminobenzonitrile, p-cyanoaniline, benzonitrile, 4-amino, aniline, p-cyano, benzonitrile, p-amino, 1-amino-4-cyanobenzene, unii-pb8v69y76t, 4-amino benzonitrile, 4-amino-benzonitrile
YBAZINRZQSAIAY-UHFFFAOYSA-N
4-aminobenzonitrile
13396
Crystalline Powder Chunks
Authentic
Beige to Orange-Yellow or White
10g
97.5
97.5% min. (GC)
H2NC6H4CN
14, 425
Solubility in water: insoluble
C1=CC(=CC=C1C#N)N
118.14
118.14
98%
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Aminobenzonitrile

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Product Certifications