missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzo-2,1,3-thiadiazole, 98%, Thermo Scientific Chemicals

Catalog Number p-7045171
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
767-64-6
C6H5N3S
151.187
MFCD00005810
DRLGIZIAMHIQHL-UHFFFAOYSA-N
4-aminobenzo-2,1,3-thiadiazole, 4-amino-2,1,3-benzothiadiazole, 4-aminopiazthiole, benzo c 1,2,5 thiadiazol-4-amine, 2,1,3-benzothiadiazole, 4-amino, 7-amino-2,1,3-benzothiadiazole, benzo 1,2,5 thiadiazol-4-ylamine, 2,1,3-benzothiadiazole, 7-amino, 2,1,3-benzothiadiazol-4-ylamine, 2,1,3-benzothiadiazol-4 1h-imine
69845
2,1,3-benzothiadiazol-4-amine
C1=CC2=NSN=C2C(=C1)N
This item is not returnable. View return policy
767-64-6
C6H5N3S
151.187
MFCD00005810
DRLGIZIAMHIQHL-UHFFFAOYSA-N
4-aminobenzo-2,1,3-thiadiazole, 4-amino-2,1,3-benzothiadiazole, 4-aminopiazthiole, benzo c 1,2,5 thiadiazol-4-amine, 2,1,3-benzothiadiazole, 4-amino, 7-amino-2,1,3-benzothiadiazole, benzo 1,2,5 thiadiazol-4-ylamine, 2,1,3-benzothiadiazole, 7-amino, 2,1,3-benzothiadiazol-4-ylamine, 2,1,3-benzothiadiazol-4 1h-imine
69845
2,1,3-benzothiadiazol-4-amine
C1=CC2=NSN=C2C(=C1)N

4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 67°C to 69°C
Quantity 1 g
Beilstein 3551
Solubility Information Insoluble in water.
Formula Weight 151.19
Percent Purity 98%
Chemical Name or Material 4-Aminobenzo-2,1,3-thiadiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'