missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminoantipyrine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
4-Aminoantipyrine shows both analgesic and anti-inflammatory properties. It is used as a reagent for glucose determination in the presence of peroxidase and phenol. It is used as a drug dipyrone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Aminoantipyrine shows both analgesic and anti-inflammatory properties. It is used as a reagent for glucose determination in the presence of peroxidase and phenol. It is used as a drug dipyrone.
Solubility
Soluble in water. 500 g/L (20°C), methanol. Partially soluble in diethyl ether.
Notes
Incompatible with Strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
4-Aminoantipyrine shows both analgesic and anti-inflammatory properties. It is used as a reagent for glucose determination in the presence of peroxidase and phenol. It is used as a drug dipyrone.
Solubility
Soluble in water. 500 g/L (20°C), methanol. Partially soluble in diethyl ether.
Notes
Incompatible with Strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
Specifications
Melting Point | 106°C to 110°C |
Assay Percent Range | 97% |
Quantity | 100 g |
Beilstein | 181635 |
Merck Index | 14,591 |
Solubility Information | Soluble in water. 500g/L (20°C),methanol. Partially soluble in diethyl ether. |
Formula Weight | 203.25 |
Percent Purity | 97% |
Chemical Name or Material | 4-Aminoantipyrine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'