missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-m-cresol, 99+%, ACROS Organics™

£36.20 - £121.00

Chemical Identifiers

CAS 2835-99-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007871
InChI Key QGNGOGOOPUYKMC-UHFFFAOYSA-N
Synonym 4-amino-m-cresol, 4-hydroxy-2-methylaniline, 3-methyl-4-aminophenol, p-amino-m-cresol, phenol, 4-amino-3-methyl, p-hydroxy-o-toluidine, 2-methyl-4-hydroxyaniline, 2-amino-5-hydroxytoluene, m-cresol, 4-amino, 4-amino-3-cresol
PubChem CID 17819
ChEBI CHEBI:55546
IUPAC Name 4-amino-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10285780
SDS View Documents
Acros Organics
163200500
50g Glass bottle
£36.20
50g
View alternative products
Save Up to 
»
In stock, ready to ship: 2
Please to purchase this item. Need a web account? Register with us today!
10031970
SDS View Documents
Acros Organics
163202500
250g Glass bottle
£121.00
250g
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 30-09-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

2835-99-6
123.15
QGNGOGOOPUYKMC-UHFFFAOYSA-N
17819
4-amino-3-methylphenol
C7H9NO
MFCD00007871
4-amino-m-cresol, 4-hydroxy-2-methylaniline, 3-methyl-4-aminophenol, p-amino-m-cresol, phenol, 4-amino-3-methyl, p-hydroxy-o-toluidine, 2-methyl-4-hydroxyaniline, 2-amino-5-hydroxytoluene, m-cresol, 4-amino, 4-amino-3-cresol
CHEBI:55546
CC1=C(C=CC(=C1)O)N

Specifications

4-Amino-m-cresol
Glass bottle
176°C to 180°C
99+%
99.0
99% min. (GC)
H2NC6H3(CH3)OH
13, 593
Solubility in water: insoluble
CC1=C(C=CC(=C1)O)N
123.15
CHEBI:55546
Powder
Authentic
Brown
50g
2835-99-6
100.0
C7H9NO
MFCD00007871
4-amino-m-cresol, 4-hydroxy-2-methylaniline, 3-methyl-4-aminophenol, p-amino-m-cresol, phenol, 4-amino-3-methyl, p-hydroxy-o-toluidine, 2-methyl-4-hydroxyaniline, 2-amino-5-hydroxytoluene, m-cresol, 4-amino, 4-amino-3-cresol
QGNGOGOOPUYKMC-UHFFFAOYSA-N
4-amino-3-methylphenol
17819
123.15
99+%
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Amino-m-cresol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Certificates