missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-8-(trifluoromethyl)quinoline, 97%, Thermo Scientific™

Catalog Number p-7033157
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
243977-15-3
C10H7F3N2
212.18
MFCD00269346
JTMLOHYYNHRKTF-UHFFFAOYSA-N
8-trifluoromethyl quinolin-4-amine, 4-amino-8-trifluoromethyl quinoline, 4-amino-8-trifluoromethylquinoline, 8-trifluoromethyl-4-quinolinamine, 8-trifluoromethyl-4-quinolylamine, 8-trifluoromethyl-quinolin-4-ylamine, 8-trifluoromethyl quinolin-4-ylamine, acmc-20ai5g, 4-quinolinamine,8-trifluoromethyl
2735961
8-(trifluoromethyl)quinolin-4-amine
NC1=CC=NC2=C(C=CC=C12)C(F)(F)F
This item is not returnable. View return policy
243977-15-3
C10H7F3N2
212.18
MFCD00269346
JTMLOHYYNHRKTF-UHFFFAOYSA-N
8-trifluoromethyl quinolin-4-amine, 4-amino-8-trifluoromethyl quinoline, 4-amino-8-trifluoromethylquinoline, 8-trifluoromethyl-4-quinolinamine, 8-trifluoromethyl-4-quinolylamine, 8-trifluoromethyl-quinolin-4-ylamine, 8-trifluoromethyl quinolin-4-ylamine, acmc-20ai5g, 4-quinolinamine,8-trifluoromethyl
2735961
8-(trifluoromethyl)quinolin-4-amine
NC1=CC=NC2=C(C=CC=C12)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Amino-8-(trifluoromethyl)quinoline is used to synthesis of rubber chemical, antiseptic, antipyretic, and antiperiodic properties and are used as antimalarials and for preparing other antimalarial drugs

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 212.18
Percent Purity 97%
Chemical Name or Material 4-Amino-8-(trifluoromethyl)quinoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'