missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-methoxypyrimidine, 97%, Thermo Scientific Chemicals

Catalog Number p-4490333
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
696-45-7
C5H7N3O
125.13
MFCD00129983
VELRBZDRGTVGGT-UHFFFAOYSA-N
4-amino-6-methoxypyrimidine, 6-methoxy-pyrimidin-4-ylamine, 6-methoxy-4-pyrimidinamine, 4-amino-6-methoxy pyrimidine, 4-pyrimidinamine, 6-methoxy, 6-methoxypyrimidine-4-ylamine, 6-methoxypyrimidin-4-amin, pubchem2605, acmc-209oa2, 6-amino-4-methoxypyrimidine
735731
6-methoxypyrimidin-4-amine
COC1=NC=NC(=C1)N
This item is not returnable. View return policy
696-45-7
C5H7N3O
125.13
MFCD00129983
VELRBZDRGTVGGT-UHFFFAOYSA-N
4-amino-6-methoxypyrimidine, 6-methoxy-pyrimidin-4-ylamine, 6-methoxy-4-pyrimidinamine, 4-amino-6-methoxy pyrimidine, 4-pyrimidinamine, 6-methoxy, 6-methoxypyrimidine-4-ylamine, 6-methoxypyrimidin-4-amin, pubchem2605, acmc-209oa2, 6-amino-4-methoxypyrimidine
735731
6-methoxypyrimidin-4-amine
COC1=NC=NC(=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 154.0°C to 159.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 125.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-6-methoxypyrimidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'