missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-6-hydroxypyrimidine, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 262.0°C to 264.0°C |
Loss on Drying | 1% max. (105°C, 3 hr) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Solubility Information | Solubility in water: slightly soluble. |
IUPAC Name | 6-amino-1H-pyrimidin-4-one |
Formula Weight | 111.1 |
Percent Purity | 98% |
Show More |
Safety and Handling
Product Identifier
- 4-Amino-6-hydroxypyrimidine
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'