missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-chloro-2-methoxybenzoic Acid 98+%, ACROS Organics™

Brand:  Acros Organics 133670250

Code : 29

Additional Details : CAS Number : 7206-70-4 Weight : 0.08000kg


Quantity 25g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10143330

  • £48.30 / 25g


Estimated Shipment
19-03-2018
Add to basket


Specifications

Chemical Identifiers

4-Amino-5-chloro-2-methoxybenzoic acid
7206-70-4
100.0
98+%
NH2(Cl)C6H2(OCH3)COOH
MFCD00038794
98+%
98.0
≥98%
C8H8ClNO3
201.61

Specifications

4-Amino-5-chloro-2-methoxybenzoic acid
Plastic bottle
Authentic
100.0
206.0° to 210.0°C
25g
≥98%
C8H8ClNO3
201.61
98+%
7206-70-4
98.0
0.2% max. (K.F.)
White to slightly beige
Powder
98+%
NH2(Cl)C6H2(OCH3)COOH
MFCD00038794
Videos
SDS