missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-bromo-2-chloropyrimidine, 97%, ACROS Organics™

Brand:  Acros Organics 429380050

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 205672-25-9 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10798288

  • £391.00 / 5g


EU Stock 1
Add to basket


Specifications Description & Specifications

Chemical Identifiers

4-Amino-5-bromo-2-chloropyrimidine
96.0
≥96.0% (GC)
C4H3BrClN3
MFCD07782040
205672-25-9
100.0
97%
208.45

Specifications

4-Amino-5-bromo-2-chloropyrimidine
96.0
Authentic
Liquid
97%
Clear brown to purple
208.45
MFCD07782040
205672-25-9
100.0
Glass bottle
≥96.0% (GC)
187° to 192°C
C4H3BrClN3
5g

Safety and Handling

Product Identifier
  • 4-Amino-5-bromo-2-chloropyrimidine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents