missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, ACS, 90+%, Thermo Scientific Chemicals

Catalog Number 15486155
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
116-63-2
C10H9NO4S
239.245
MFCD00004019
RXCMFQDTWCCLBL-UHFFFAOYSA-N
1-amino-2-naphthol-4-sulfonic acid, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 1-amino-4-sulfo-2-naphthol, 1,2,4-acid, unii-w46sk3p33g, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, 1,2,4-aminonaphtholsulfonic acid, 2-hydroxy-4-sulfo-1-naphthylamine
8316
CHEBI:19024
4-amino-3-hydroxynaphthalene-1-sulfonic acid
C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
This item is not returnable. View return policy
116-63-2
C10H9NO4S
239.245
MFCD00004019
RXCMFQDTWCCLBL-UHFFFAOYSA-N
1-amino-2-naphthol-4-sulfonic acid, 4-amino-3-hydroxy-1-naphthalenesulfonic acid, 1-amino-4-sulfo-2-naphthol, 1,2,4-acid, unii-w46sk3p33g, 1-naphthalenesulfonic acid, 4-amino-3-hydroxy, 4-amino-3-hydroxynaphthalene-1-sulphonic acid, 4-amino-3-hydroxynaphthalenesulfonic acid, 1,2,4-aminonaphtholsulfonic acid, 2-hydroxy-4-sulfo-1-naphthylamine
8316
CHEBI:19024
4-amino-3-hydroxynaphthalene-1-sulfonic acid
C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O

4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as a key starting material for the preparation of naphthalenesulfonic acid derivatives by reacting with palmitoyl chloride, which shows the activity to inhibit DNA polymerase and ribonuclease H Activities of HIV-1 reverse transcriptase. It is also used to prepare a copolyner with melamine, which is associated with graphene to prepare molecularly imprinted sensor and utilized for the determination of melatonin in human biological fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼295°C (decomposition)
Quantity 5 g
Beilstein 2697469
Merck Index 14,454
Solubility Information Soluble in hot water,hot hydrogen sulfite of sodium and alkali. Insoluble in water,ethanol,ethyl ether and benzene.
Formula Weight 239.25
Percent Purity ≥98%
Grade ACS
Physical Form Powder
Chemical Name or Material 4-Amino-3-hydroxy-1-naphthalenesulfonic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'