Learn More
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, ACS, 90+%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as a key starting material for the preparation of naphthalenesulfonic acid derivatives by reacting with palmitoyl chloride, which shows the activity to inhibit DNA polymerase and ribonuclease H Activities of HIV-1 reverse transcriptase. It is also used to prepare a copolyner with melamine, which is associated with graphene to prepare molecularly imprinted sensor and utilized for the determination of melatonin in human biological fluids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼295°C (decomposition) |
Quantity | 100 g |
Beilstein | 2697469 |
Merck Index | 14,454 |
Solubility Information | Soluble in hot water,hot hydrogen sulfite of sodium and alkali. Insoluble in water,ethanol,ethyl ether and benzene. |
Formula Weight | 239.25 |
Percent Purity | ≥98% |
Grade | ACS |
Physical Form | Powder |
Chemical Name or Material | 4-Amino-3-hydroxy-1-naphthalenesulfonic acid |
RUO – Research Use Only