missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 99%, Thermo Scientific Chemicals
126.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as mordant dyes and acid dyes intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as mordant dyes and acid dyes intermediate.
Solubility
Soluble in bases. Insoluble in water.
Notes
Store away from strong oxidizers, strong bases, acid anhydrides, acid chlorides.
4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as mordant dyes and acid dyes intermediate.
Solubility
Soluble in bases. Insoluble in water.
Notes
Store away from strong oxidizers, strong bases, acid anhydrides, acid chlorides.
Specifications
Melting Point | ∼295°C (decomposition) |
Quantity | 100 g |
Beilstein | 2697469 |
Merck Index | 14,454 |
Solubility Information | Soluble in bases. Insoluble in water. |
Formula Weight | 239.25 |
Percent Purity | 99% |
Chemical Name or Material | 4-Amino-3-hydroxy-1-naphthalenesulfonic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'