missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-bromopyridine, 98%, ACROS Organics™

£37.57 - £129.00

Chemical Identifiers

CAS 13534-98-0
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.01
MDL Number MFCD02068297
InChI Key DDQYSZWFFXOXER-UHFFFAOYSA-N
Synonym 4-amino-3-bromopyridine, 3-bromo-4-pyridinamine, 3-bromo-4-aminopyridine, 3-bromo-4-pyridineamine, 4-pyridinamine, 3-bromo, 3-bromo-4-pyridylamine, 3-bromo-4-pyridinylamine, 3-bromo-pyridin-4-ylamine, pubchem1242, acmc-1bnz6
PubChem CID 26095
IUPAC Name 3-bromopyridin-4-amine
SMILES C1=CN=CC(=C1N)Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10571632
SDS View Documents
Acros Organics
360620010
1g Glass bottle
£37.57
1g
View alternative products
Save Up to 
»
Estimated Shipment
08-12-2020
Please to purchase this item. Need a web account? Register with us today!
10025593
SDS View Documents
Acros Organics
360620050
5g Glass bottle
£129.00
5g
View alternative products
Save Up to 
»
Estimated Shipment
08-12-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

13534-98-0
173.01
DDQYSZWFFXOXER-UHFFFAOYSA-N
26095
C1=CN=CC(=C1N)Br
C5H5BrN2
MFCD02068297
4-amino-3-bromopyridine, 3-bromo-4-pyridinamine, 3-bromo-4-aminopyridine, 3-bromo-4-pyridineamine, 4-pyridinamine, 3-bromo, 3-bromo-4-pyridylamine, 3-bromo-4-pyridinylamine, 3-bromo-pyridin-4-ylamine, pubchem1242, acmc-1bnz6
3-bromopyridin-4-amine

Specifications

4-Amino-3-bromopyridine
Glass bottle
67.0°C to 71.0°C
13534-98-0
100.0
C5H5BrN2
22, V,9, 185
DDQYSZWFFXOXER-UHFFFAOYSA-N
3-bromopyridin-4-amine
26095
Crystalline Powder
Authentic
White
1g
97.5
97.5% min. (GC)
MFCD02068297
4-amino-3-bromopyridine, 3-bromo-4-pyridinamine, 3-bromo-4-aminopyridine, 3-bromo-4-pyridineamine, 4-pyridinamine, 3-bromo, 3-bromo-4-pyridylamine, 3-bromo-4-pyridinylamine, 3-bromo-pyridin-4-ylamine, pubchem1242, acmc-1bnz6
C1=CN=CC(=C1N)Br
173.01
173.01
98%
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Amino-3-bromopyridine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS