missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-bromopyridine, 98%, ACROS Organics™

Brand:  Acros Organics 360620010

Code : 29

Additional Details : CAS Number : 13534-98-0 Weight : 0.05100kg


Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 10571632

  • £35.50 / 1g

Estimated Shipment date
from Supplier 04-03-2020

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

13534-98-0
173.01
DDQYSZWFFXOXER-UHFFFAOYSA-N
26095
C1=CN=CC(=C1N)Br
C5H5BrN2
MFCD02068297
4-amino-3-bromopyridine, 3-bromo-4-pyridinamine, 3-bromo-4-aminopyridine, 3-bromo-4-pyridineamine, 4-pyridinamine, 3-bromo, 3-bromo-4-pyridylamine, 3-bromo-4-pyridinylamine, 3-bromo-pyridin-4-ylamine, pubchem1242, acmc-1bnz6
3-bromopyridin-4-amine

Specifications

4-Amino-3-bromopyridine
Glass bottle
67.0°C to 71.0°C
13534-98-0
100.0
C5H5BrN2
22, V,9, 185
DDQYSZWFFXOXER-UHFFFAOYSA-N
3-bromopyridin-4-amine
26095
Crystalline Powder
Authentic
White
1g
97.5
97.5% min. (GC)
MFCD02068297
4-amino-3-bromopyridine, 3-bromo-4-pyridinamine, 3-bromo-4-aminopyridine, 3-bromo-4-pyridineamine, 4-pyridinamine, 3-bromo, 3-bromo-4-pyridylamine, 3-bromo-4-pyridinylamine, 3-bromo-pyridin-4-ylamine, pubchem1242, acmc-1bnz6
C1=CN=CC(=C1N)Br
173.01
173.01
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Amino-3-bromopyridine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents