missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-diiodobenzoic acid, tech. 90%, Thermo Scientific Chemicals

Catalog Number p-7080881
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2122-61-4
C7H5I2NO2
388.93
MFCD00007684
WXTVPMWCUMEVSZ-UHFFFAOYSA-N
benzoic acid, 4-amino-3,5-diiodo, 3,5-dijod-4-aminohippursaeure, 3,5-diiodo-4-aminobenzoic acid, 3,5-dijod-4-aminohippursaeure german, 4-amino-3,5-diiodo-benzoicacid, pubchem4681, acmc-1cq3z, wln: zr bi fi dvq, labotest-bb lt00080707, 4-amino-3,5-diiodo-benzoicaci
16460
4-amino-3,5-diiodobenzoic acid
NC1=C(I)C=C(C=C1I)C(O)=O
This item is not returnable. View return policy
2122-61-4
C7H5I2NO2
388.93
MFCD00007684
WXTVPMWCUMEVSZ-UHFFFAOYSA-N
benzoic acid, 4-amino-3,5-diiodo, 3,5-dijod-4-aminohippursaeure, 3,5-diiodo-4-aminobenzoic acid, 3,5-dijod-4-aminohippursaeure german, 4-amino-3,5-diiodo-benzoicacid, pubchem4681, acmc-1cq3z, wln: zr bi fi dvq, labotest-bb lt00080707, 4-amino-3,5-diiodo-benzoicaci
16460
4-amino-3,5-diiodobenzoic acid
NC1=C(I)C=C(C=C1I)C(O)=O

4-Amino-3,5-diiodobenzoic acid is a very useful substituted benzene compound for use as a building block in synthesis. The carboxylic acid and amine functionalities can be used to form linkages through nucleophile-electrophile coupling and the iodides may be used as functional sites for cross-coupling transformations. 4-Amino-3,5-diiodobenzoic acid has been specifically used in the synthesis of an adduct to introduce into Tecoflex polymers to impart radiopaqueness to the material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3,5-diiodobenzoic acid is a very useful substituted benzene compound for use as a building block in synthesis. The carboxylic acid and amine functionalities can be used to form linkages through nucleophile-electrophile coupling and the iodides may be used as functional sites for cross-coupling transformations. 4-Amino-3,5-diiodobenzoic acid has been specifically used in the synthesis of an adduct to introduce into Tecoflex polymers to impart radiopaqueness to the material.

Solubility
Soluble in water (partly miscible).

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 5 g
Beilstein 2103037
Sensitivity Light sensitive
Solubility Information Soluble in water (partly miscible).
Formula Weight 388.93
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Amino-3,5-diiodobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'