missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number 15497686
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
939968-08-8
C12H16BF2NO2
255.072
MFCD18434474
JGOZEXIYNJERIP-UHFFFAOYSA-N
2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3,5-difluorophenyl boronic acid pinacol ester, 4-amino-3,5-difluorobenzeneboronic acid pinacol ester, 4-amino-3,5-difluorophenylboronic acid pinacol ester, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxabor, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
59231931
2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)F)N)F
This item is not returnable. View return policy
939968-08-8
C12H16BF2NO2
255.072
MFCD18434474
JGOZEXIYNJERIP-UHFFFAOYSA-N
2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3,5-difluorophenyl boronic acid pinacol ester, 4-amino-3,5-difluorobenzeneboronic acid pinacol ester, 4-amino-3,5-difluorophenylboronic acid pinacol ester, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxabor, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
59231931
2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)F)N)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Formula Weight 255.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Amino-3,5-difluorobenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'