missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-dichlorobenzoic acid, 98+%, Thermo Scientific™

Catalog Number p-3593315
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
56961-25-2
C7H5Cl2NO2
206.02
MFCD00017091
UHXYYTSWBYTDPD-UHFFFAOYSA-N
3,5-dichloro-4-aminobenzoic acid, unii-bik7ns3b2f, benzoic acid, 4-amino-3,5-dichloro, bik7ns3b2f, 4-amino-3,5-dichlorobenzoicacid, pubchem14451, acmc-209xmx, 4-14-00-01279 beilstein handbook reference, ksc495s0n, 4-carboxy-2,6-dichloroaniline
42061
4-amino-3,5-dichlorobenzoic acid
NC1=C(Cl)C=C(C=C1Cl)C(O)=O
This item is not returnable. View return policy
56961-25-2
C7H5Cl2NO2
206.02
MFCD00017091
UHXYYTSWBYTDPD-UHFFFAOYSA-N
3,5-dichloro-4-aminobenzoic acid, unii-bik7ns3b2f, benzoic acid, 4-amino-3,5-dichloro, bik7ns3b2f, 4-amino-3,5-dichlorobenzoicacid, pubchem14451, acmc-209xmx, 4-14-00-01279 beilstein handbook reference, ksc495s0n, 4-carboxy-2,6-dichloroaniline
42061
4-amino-3,5-dichlorobenzoic acid
NC1=C(Cl)C=C(C=C1Cl)C(O)=O
CAS Min % 98.0
CAS Max % 100.0
Melting Point 288°C to 290°C
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula NH2C6H2(Cl)2CO2H
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 206.03
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 4-Amino-3, 5-dichlorobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-3,5-dichlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'