missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number 15465366
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1326283-60-6
C13H19BFNO3
267.11
MFCD22419265
JWACNMHKJHEWAQ-UHFFFAOYSA-N
4-amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 5-fluoro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline
67363295
5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
COC1=C(N)C=C(F)C(=C1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
1326283-60-6
C13H19BFNO3
267.11
MFCD22419265
JWACNMHKJHEWAQ-UHFFFAOYSA-N
4-amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 5-fluoro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline
67363295
5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
COC1=C(N)C=C(F)C(=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 267.11
Percent Purity 96%
Chemical Name or Material 4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'