missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloro-3-iodopyridine, 95%, Thermo Scientific™

Catalog Number 15435937
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
909036-46-0
C5H4ClIN2
254.46
MFCD09763660
BYVMKCHHWASQFN-UHFFFAOYSA-N
2-chloro-3-iodo-4-pyridinamine, 2-chloro-3-iodo-4-aminopyridine, 4-amino-2-chloro-3-iodopyridine, pubchem22625, 2-chloro-3-iodo-4-pyridylamine, 2-chloro-3-iodo-pyridin-4-amine, 2-chloro-3-iodopyridin-4-ylamine, 2-chloro-3-iodo-pyridin-4-ylamine, 4-pyridinamine, 2-chloro-3-iodo
24229208
2-chloro-3-iodopyridin-4-amine
NC1=C(I)C(Cl)=NC=C1
This item is not returnable. View return policy
909036-46-0
C5H4ClIN2
254.46
MFCD09763660
BYVMKCHHWASQFN-UHFFFAOYSA-N
2-chloro-3-iodo-4-pyridinamine, 2-chloro-3-iodo-4-aminopyridine, 4-amino-2-chloro-3-iodopyridine, pubchem22625, 2-chloro-3-iodo-4-pyridylamine, 2-chloro-3-iodo-pyridin-4-amine, 2-chloro-3-iodopyridin-4-ylamine, 2-chloro-3-iodo-pyridin-4-ylamine, 4-pyridinamine, 2-chloro-3-iodo
24229208
2-chloro-3-iodopyridin-4-amine
NC1=C(I)C(Cl)=NC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 116°C to 117°C
Color Orange-Pink to White
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 254.46
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Amino-2-chloro-3-iodopyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'