missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dihydroxypyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number 10598460
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
873-83-6
C4H5N3O2
127.10
MFCD00006071
LNDZXOWGUAIUBG-UHFFFAOYSA-N
6-aminouracil, 4-amino-2,6-dihydroxypyrimidine, 6-aminopyrimidine-2,4 1h,3h-dione, 4-aminouracil, 6-aminopyrimidine-2,4-diol, 6-amino-2,4-pyrimidinediol, 2,4 1h,3h-pyrimidinedione, 6-amino, uracil, 6-amino, cytosine, 6-hydroxy, 6-amino-2,4-dihydroxypyrimidine
70120
NC1=CC(=O)NC(=O)N1
This item is not returnable. View return policy
873-83-6
C4H5N3O2
127.10
MFCD00006071
LNDZXOWGUAIUBG-UHFFFAOYSA-N
6-aminouracil, 4-amino-2,6-dihydroxypyrimidine, 6-aminopyrimidine-2,4 1h,3h-dione, 4-aminouracil, 6-aminopyrimidine-2,4-diol, 6-amino-2,4-pyrimidinediol, 2,4 1h,3h-pyrimidinedione, 6-amino, uracil, 6-amino, cytosine, 6-hydroxy, 6-amino-2,4-dihydroxypyrimidine
70120
NC1=CC(=O)NC(=O)N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Brown to Cream
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 24, 469
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in methanol
IUPAC Name 6-amino-1H-pyrimidine-2,4-dione
Formula Weight 127.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2, 6-dihydroxypyrimidine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-2,6-dihydroxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'