missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dichlorophenol, 98%, Thermo Scientific Chemicals

Catalog Number p-55682
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5930-28-9
C6H5Cl2NO
178.01
MFCD00007875
KGEXISHTCZHGFT-UHFFFAOYSA-N
2,6-dichloro-4-aminophenol, phenol, 4-amino-2,6-dichloro, 2,6-dichloro-p-aminophenol, 3,5-dichloro-4-hydroxyaniline, unii-i0l4csx5n0, i0l4csx5n0, 2,6-dichloro-4-amino phenol, 4-amino-2,6-dichloro-phenol, acmc-209mc4, dsstox_cid_20720
80037
4-amino-2,6-dichlorophenol
NC1=CC(Cl)=C(O)C(Cl)=C1
This item is not returnable. View return policy
5930-28-9
C6H5Cl2NO
178.01
MFCD00007875
KGEXISHTCZHGFT-UHFFFAOYSA-N
2,6-dichloro-4-aminophenol, phenol, 4-amino-2,6-dichloro, 2,6-dichloro-p-aminophenol, 3,5-dichloro-4-hydroxyaniline, unii-i0l4csx5n0, i0l4csx5n0, 2,6-dichloro-4-amino phenol, 4-amino-2,6-dichloro-phenol, acmc-209mc4, dsstox_cid_20720
80037
4-amino-2,6-dichlorophenol
NC1=CC(Cl)=C(O)C(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 164°C to 168°C
Color Beige or Brown to Red
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC6H2(Cl)2OH
Quantity 5 g
Beilstein 13, 512
Solubility Information (2.5% in 96% ethanol) Clear to slightly hazy
Formula Weight 178.02
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Amino-2, 6-dichlorophenol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-2,6-dichlorophenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'