missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,5-dimethylphenol, 95%, Thermo Scientific™

Catalog Number p-3756781
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3096-71-7
C8H11NO
137.18
MFCD00007873
JSWVCUXQICMATE-UHFFFAOYSA-N
4-amino-2,5-xylenol, phenol, 4-amino-2,5-dimethyl, unii-rf4r93709t, 2,5-dimethyl-4-aminophenol, 4-amino-2,5-dimethyl-phenol, 4-hydroxy-2,5-dimethylaniline, acmc-209hjo, 2,5-dimethy-4-aminophenol, 2,5-xylenol, 4-amino, ksc495q0r
76544
4-amino-2,5-dimethylphenol
CC1=CC(=C(C=C1O)C)N
This item is not returnable. View return policy
3096-71-7
C8H11NO
137.18
MFCD00007873
JSWVCUXQICMATE-UHFFFAOYSA-N
4-amino-2,5-xylenol, phenol, 4-amino-2,5-dimethyl, unii-rf4r93709t, 2,5-dimethyl-4-aminophenol, 4-amino-2,5-dimethyl-phenol, 4-hydroxy-2,5-dimethylaniline, acmc-209hjo, 2,5-dimethy-4-aminophenol, 2,5-xylenol, 4-amino, ksc495q0r
76544
4-amino-2,5-dimethylphenol
CC1=CC(=C(C=C1O)C)N
CAS Min % 94.0
CAS Max % 100.0
Melting Point 242°C to 243°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2C6H2(OH)NH2
Quantity 5 g
Beilstein 13, 634
Formula Weight 137.18
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Amino-2, 5-dimethylphenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-2,5-dimethylphenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'