missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.
Notes
Air sensitive. Store away from oxidizing agents.
4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.
Notes
Air sensitive. Store away from oxidizing agents.
Specifications
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Formula Weight | 255.07 |
Percent Purity | 96% |
Chemical Name or Material | 4-Amino-2,5-difluorobenzeneboronic acid pinacol ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'