missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,2,6,6-tetramethylpiperidinooxy, 97%, free radical, Thermo Scientific Chemicals

Catalog Number 10685042
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
14691-88-4
C9H19N2O
171.26
MFCD00006479
4-Amino-TEMPO
This item is not returnable. View return policy
14691-88-4
C9H19N2O
171.26
MFCD00006479
4-Amino-TEMPO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Polymerization reagent
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,chloroform,hexane
Formula Weight 171.26
Percent Purity 97%
Physical Form Adhering Crystals, Needles or Moist Mass
Chemical Name or Material 4-Amino-2,2,6,6-tetramethylpiperidinooxy
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Amino-2,2,6,6-tetramethylpiperidinooxy
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'