missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine, tech. 85%, Thermo Scientific™

Catalog Number p-7044563
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
5334-30-5
C11H9N5
211.23
MFCD00022901
KKDPIZPUTYIBFX-UHFFFAOYSA-N
4879
NC1=C2C=NN(C2=NC=N1)C1=CC=CC=C1
This item is not returnable. View return policy
5334-30-5
C11H9N5
211.23
MFCD00022901
KKDPIZPUTYIBFX-UHFFFAOYSA-N
4879
NC1=C2C=NN(C2=NC=N1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 214°C to 217°C
Quantity 1 g
Beilstein 198460
Synonym 4-amino-1-phenylpyrazolo 3,4-d pyrimidine, 1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4-amine, pp3, pp 3, 1-phenylpyrazolo 3,4-d pyrimidin-4-amine, chembl65063, 1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4-ylamine, 4-amino-7-phenylpyrazol 3,4-d pyrimidine, 4-amino-1-phenylpyrazolo 3,4-e pyrimidine, 1h-pyrazolo 3,4-d pyrimidin-4-amine, 1-phenyl
IUPAC Name 1-phenylpyrazolo[3,4-d]pyrimidin-4-amine
Formula Weight 211.23
Percent Purity 85%
Grade Technical
Chemical Name or Material 4-Amino-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'