missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Amino-1-benzylpiperidine, 98%

Brand:  Alfa Aesar™ A10149.06

Code : NEW

Additional Details : CAS Number : 50541-93-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11419422

  • £26.00 / 5g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Amino-1-benzylpiperidine
C12H18N2
MFCD00006504
1-Benzyl-4-pyridinamine
50541-93-0
190.29
146038

Specifications

4-Amino-1-benzylpiperidine
98%
279° to 281°C
5g
MFCD00006504
1-Benzyl-4-pyridinamine
50541-93-0
C12H18N2
190.29
>110°C (230°F)
146038
Videos
SDS