missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylpyridine, 98%, ACROS Organics™

Brand:  Acros Organics 102705000

Code : 29

Additional Details : CAS Number : 1122-54-9 Weight : 0.55000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III


Packaging Glass bottle
Quantity 500g

 View more versions of this product

Product Code. 10474545

  • £195.40 / 500g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

4-Acetylpyridine
97.5
98%
C7H7NO
121.14
Methyle4-pyridyl ketone
1122-54-9
100.0
98%
(C5H4N)COCH3
MFCD00006433

Specifications

4-Acetylpyridine
97.5
Authentic
98%
C7H7NO
(C5H4N)COCH3
MFCD00006433
21, 279
1.09
Clear light yellow to brownish
212.0°C
104°C
1122-54-9
100.0
Liquid
98%
Glass bottle
121.14
1.5270 to 1.5310
Methyle4-pyridyl ketone
1.0900g/mL
13.0° to 16.0°C
500g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Acetylpyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS