missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-4-phenylpiperidine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number 10582531
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10315-03-4
C13H17NO·ClH
239.74
MFCD00039037
JYDHZOIDIWUHDB-UHFFFAOYSA-N
4-acetyl-4-phenylpiperidine hydrochloride, 1-4-phenylpiperidin-4-yl ethanone hydrochloride, 1-4-phenylpiperidin-4-yl ethan-1-one hydrochloride, 4-acetyl-4-phenylpiperidine, chloride, 1-4-phenylpiperidin-4-yl ethan-1-one hcl, acmc-1c3wz, 4-acetyl-4-phenylpiperidine hcl, 4-acetyl 4-phenyl piperidine hydrochloride, 4-acetyl-4-phenyl-piperidine hydrochloride
2723767
1-(4-phenylpiperidin-4-yl)ethanone;hydrochloride
CC(=O)C1(CCNCC1)C2=CC=CC=C2.Cl
This item is not returnable. View return policy
10315-03-4
C13H17NO·ClH
239.74
MFCD00039037
JYDHZOIDIWUHDB-UHFFFAOYSA-N
4-acetyl-4-phenylpiperidine hydrochloride, 1-4-phenylpiperidin-4-yl ethanone hydrochloride, 1-4-phenylpiperidin-4-yl ethan-1-one hydrochloride, 4-acetyl-4-phenylpiperidine, chloride, 1-4-phenylpiperidin-4-yl ethan-1-one hcl, acmc-1c3wz, 4-acetyl-4-phenylpiperidine hcl, 4-acetyl 4-phenyl piperidine hydrochloride, 4-acetyl-4-phenyl-piperidine hydrochloride
2723767
1-(4-phenylpiperidin-4-yl)ethanone;hydrochloride
CC(=O)C1(CCNCC1)C2=CC=CC=C2.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 232°C to 234°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Formula Weight 239.74
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Acetyl-4-phenylpiperidine hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'