missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number p-7054332
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1351499-39-2
C14H18BFO3
264.103
UDNSDVWJDRQRHR-UHFFFAOYSA-N
1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-acetyl-3-fluorophenylboronic acid pinacol ester
66802112
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(=O)C)F
This item is not returnable. View return policy
1351499-39-2
C14H18BFO3
264.103
UDNSDVWJDRQRHR-UHFFFAOYSA-N
1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-acetyl-3-fluorophenylboronic acid pinacol ester
66802112
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(=O)C)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 264.1
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Acetyl-3-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'