missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Acetyl-2-fluorobenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62256.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447216

  • £330.10 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Acetyl-2-fluorobenzeneboronic acid pinacol ester
264.1
4-Acetyl-2-fluorophenylboronic acid pinacol ester

Specifications

4-Acetyl-2-fluorobenzeneboronic acid pinacol ester
96%
4-Acetyl-2-fluorophenylboronic acid pinacol ester
Solid
264.1
1g
Videos
SDS