missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidothiophenol 95%, ACROS Organics™

Brand:  Acros Organics 102340250

Code : 29

Additional Details : CAS Number : 1126-81-4 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10457280

  • £121.40 / 25g


Estimated Shipment
21-05-2018
Add to basket


Specifications

Chemical Identifiers

4-Acetamidothiophenol
1126-81-4
100.0
95%
CH3CONHC6H4SH
MFCD00004848
95%
94.0
≥94%
C8H9NOS
167.23
p-Mercaptoacetanilide

Specifications

4-Acetamidothiophenol
1126-81-4
100.0
Powder
95%
CH3CONHC6H4SH
MFCD00004848
13, 541
Yellow
25g
95%
94.0
Authentic
≥94%
C8H9NOS
167.23
Glass bottle
p-Mercaptoacetanilide
149° to 154°C
Videos
SDS