missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetamidophenol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 168.0°C to 172.0°C |
Color | White |
Density | 1.2930g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Plastic Drum |
Linear Formula | CH3CONHC6H4OH |
Quantity | 5 kg |
Show More |
Safety and Handling
Product Identifier
- 4-Acetamidophenol
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'