missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Acetamidobenzenesulfonyl chloride, 98+%

Brand:  Alfa Aesar™ A11511.18

Code : NEW

Additional Details : CAS Number : 121-60-8 Weight : 0.05000kg Transport : UN number : 3261 Chem class : 1a Pack group : II

Quantity 50g

 View more versions of this product

Product Code. 11369439

  • £9.60 / 50g


Estimated Shipment date
from Supplier 27-04-2018
Add to basket

Description

Description

Applications
N-Acetylsulfanilyl chloride is used in the preparation of sulfanilamide and its derivatives which are intermediates to produce sulfa drugs. They are used in the prevention and treatment of bacterial infections, diabetes mellitus, edema, hypertension, and gout. It is also used as a Pharma raw material. N-Acetylsulfanilyl chloride widely used in the fields of dye, medicine, mainly used for preparation of Sulfanilamide, Sulfanilylureal, Sulfatolamide, Sulphathiourea , Sulfaguanidine and Sulfacetamide and etc.

Solubility
It is slightly in water. But soluble in benzene, chloroform and ether.

Notes
Stable under ordinary conditions. Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents, Strong bases.
Specifications

Chemical Identifiers

4-Acetamidobenzenesulfonyl chloride
C8H8ClNO3S
MFCD00007442
746676
N-Acetylsulfanilyl chloride
121-60-8
233.67
UN3261
14,103

Specifications

4-Acetamidobenzenesulfonyl chloride
≥98%
233.67
UN3261
14,103
≈146°C (dec.)
121-60-8
C8H8ClNO3S
MFCD00007442
746676
N-Acetylsulfanilyl chloride
50g
Videos
SDS