missing translation for 'onlineSavingsMsg'
Learn More

4-(9-Carbazolyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog Number 15492009
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
419536-33-7
C18H14BNO2
287.13
MFCD13176534
JGAVTCVHDMOQTJ-UHFFFAOYSA-N
4-9h-carbazol-9-yl phenyl boronic acid, 4-9h-carbozol-9-yl phenylboronic acid, 4-9h-carbazol-9-yl phenylboronic acid, 4-9-carbazolyl benzeneboronic acid, 4-9h-9-carbozale phenylboronic acid, 4-9-carbazolyl phenylboronic acid, 4-carbazol-9-yl phenylboronic acid, boronic acid, 4-9h-carbazol-9-yl phenyl, amtb338
23120827
OB(O)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
This item is not returnable. View return policy
419536-33-7
C18H14BNO2
287.13
MFCD13176534
JGAVTCVHDMOQTJ-UHFFFAOYSA-N
4-9h-carbazol-9-yl phenyl boronic acid, 4-9h-carbozol-9-yl phenylboronic acid, 4-9h-carbazol-9-yl phenylboronic acid, 4-9-carbazolyl benzeneboronic acid, 4-9h-9-carbozale phenylboronic acid, 4-9-carbazolyl phenylboronic acid, 4-carbazol-9-yl phenylboronic acid, boronic acid, 4-9h-carbazol-9-yl phenyl, amtb338
23120827
OB(O)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Beilstein 10565612
IUPAC Name (4-carbazol-9-ylphenyl)boronic acid
Formula Weight 287.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-(9-Carbazolyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'