missing translation for 'onlineSavingsMsg'
Learn More

4μ8C, Tocris Bioscience™

Inhibits IRE1α endoribonuclease activity

Brand:  Tocris (Bio-Techne) 447950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13464814

  • £639.97 / 50mg


Estimated Shipment
29-03-2019
Add to basket

Description

Description

Selective inhibitor of IRE1α ribonuclease (RNase) activity (IC 50 = 60 nM). Blocks substrate access to the active site of IRE1α. Inactivates IRE1α-mediated mRNA degradation. Displays no effect on the IRE1α-related endoribonuclease RNase L.
Specifications

Chemical Identifiers

14003-96-4
204.181
7-hydroxy-4-methyl-2-oxo-2h-chromene-8-carbaldehyde, 4mu8c, 8-formyl-7-hydroxy-4-methylcoumarin, 2h-1-benzopyran-8-carboxaldehyde, 7-hydroxy-4-methyl-2-oxo, 4micro8c, 8-formyl-7-hydroxy-4-methyl-coumarin, 7-hydroxy-4-methyl-2-oxo-2h-1-benzopyran-8-carbaldehyde, 7-hydroxy-4-methyl-2-oxo-2h-1-benzopyran-8-carboxaldehyde
7-hydroxy-4-methyl-2-oxochromene-8-carbaldehyde
C11H8O4
RTHHSXOVIJWFQP-UHFFFAOYSA-N
12934390
CC1=CC(=O)OC2=C1C=CC(=C2C=O)O

Specifications

4μ8C
14003-96-4
7-hydroxy-4-methyl-2-oxo-2h-chromene-8-carbaldehyde, 4mu8c, 8-formyl-7-hydroxy-4-methylcoumarin, 2h-1-benzopyran-8-carboxaldehyde, 7-hydroxy-4-methyl-2-oxo, 4micro8c, 8-formyl-7-hydroxy-4-methyl-coumarin, 7-hydroxy-4-methyl-2-oxo-2h-1-benzopyran-8-carbaldehyde, 7-hydroxy-4-methyl-2-oxo-2h-1-benzopyran-8-carboxaldehyde
CC1=CC(=O)OC2=C1C=CC(=C2C=O)O
204.181
204.18
50mg
C11H8O4
RTHHSXOVIJWFQP-UHFFFAOYSA-N
7-hydroxy-4-methyl-2-oxochromene-8-carbaldehyde
12934390
>98%
Videos