missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(7-Chloro-4-quinolinylamino)-2-(diethylaminomethyl)phenol dihydrochloride dihydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
An anti-malarial quinoline derivative that acts as an histamine N-methyltransferase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
An anti-malarial quinoline derivative that acts as an histamine N-methyltransferase inhibitor.
Specifications
Color | Yellow |
Quantity | 5 g |
Merck Index | 14,572 |
Solubility Information | Soluble in DMSO or methanol |
Formula Weight | 464.82 (392.33 Anhydrous) |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 4-(7-Chloro-4-quinolinylamino)-2-(diethylaminomethyl)phenol dihydrochloride dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'