missing translation for 'onlineSavingsMsg'
Learn More

4-[6-(Tributylstannyl)-2-pyridinyl]morpholine, 95%, Thermo Scientific™

Catalog Number 10012264
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
869901-24-6
C21H38N2OSn
453.258
MFCD08271894
LDERBIPTTICDSG-UHFFFAOYSA-N
4-6-tributylstannyl pyridin-2-yl morpholine, 4-6-tributylstannyl-2-pyridinyl morpholine, pubchem21425, 4-6-tributylstannanyl-pyridin-2-yl-morpholine, morpholine,4-6-tributylstannyl-2-pyridinyl
18525739
tributyl-(6-morpholin-4-ylpyridin-2-yl)stannane
CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)N2CCOCC2
This item is not returnable. View return policy
869901-24-6
C21H38N2OSn
453.258
MFCD08271894
LDERBIPTTICDSG-UHFFFAOYSA-N
4-6-tributylstannyl pyridin-2-yl morpholine, 4-6-tributylstannyl-2-pyridinyl morpholine, pubchem21425, 4-6-tributylstannanyl-pyridin-2-yl-morpholine, morpholine,4-6-tributylstannyl-2-pyridinyl
18525739
tributyl-(6-morpholin-4-ylpyridin-2-yl)stannane
CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)N2CCOCC2
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 453.25
Percent Purity 95%
Chemical Name or Material 4-[6-(Tributylstannyl)-2-pyridinyl]morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'