missing translation for 'onlineSavingsMsg'
Learn More
Learn More
{4-[(6-Methylpyrazin-2-yl)oxy]phenyl}methanol, 97%, Maybridge™
£31.14 - £72.33
Chemical Identifiers
CAS | 906353-02-4 |
---|---|
Molecular Formula | C12H12N2O2 |
Molecular Weight (g/mol) | 216.24 |
MDL Number | MFCD09817533 |
InChI Key | RNFIYSOVNHRFJW-UHFFFAOYSA-N |
Synonym | 4-6-methylpyrazin-2-yl oxy phenyl methanol, 4-6-methylpyrazin-2-yl oxy benzyl alcohol, 4-6-methylpyrazin-2-yloxy phenyl methan-1-ol |
PubChem CID | 24229691 |
IUPAC Name | [4-(6-methylpyrazin-2-yl)oxyphenyl]methanol |
SMILES | CC1=CN=CC(=N1)OC2=CC=C(C=C2)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10432124
|
Maybridge
CC62709CB |
250mg | Amber Glass Bottle |
£31.14
250mg |
Estimated Shipment 22-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10062504
|
Maybridge
CC62709DA |
1g | Amber Glass Bottle |
£72.33
1g |
Estimated Shipment 22-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
906353-02-4 | |
216.24 | |
RNFIYSOVNHRFJW-UHFFFAOYSA-N | |
24229691 | |
CC1=CN=CC(=N1)OC2=CC=C(C=C2)CO |
C12H12N2O2 | |
MFCD09817533 | |
4-6-methylpyrazin-2-yl oxy phenyl methanol, 4-6-methylpyrazin-2-yl oxy benzyl alcohol, 4-6-methylpyrazin-2-yloxy phenyl methan-1-ol | |
[4-(6-methylpyrazin-2-yl)oxyphenyl]methanol |
Specifications
{4-[(6-Methylpyrazin-2-yl)oxy]phenyl}methanol | |
250mg | |
96.5 | |
97% | |
MFCD09817533 | |
RNFIYSOVNHRFJW-UHFFFAOYSA-N | |
[4-(6-methylpyrazin-2-yl)oxyphenyl]methanol | |
24229691 | |
97% |
Amber Glass Bottle | |
906353-02-4 | |
100.0 | |
C12H12N2O2 | |
4-6-methylpyrazin-2-yl oxy phenyl methanol, 4-6-methylpyrazin-2-yl oxy benzyl alcohol, 4-6-methylpyrazin-2-yloxy phenyl methan-1-ol | |
CC1=CN=CC(=N1)OC2=CC=C(C=C2)CO | |
216.24 | |
216.24 |