missing translation for 'onlineSavingsMsg'
Learn More

4-(6-Methyl-2-benzimidazolyl)benzamidoxime, 97%, Thermo Scientific™

Catalog Number p-7034561
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1256486-22-2
C15H14N4O
266.304
MFCD16658868
SQIGGAWUCHCZKN-UHFFFAOYSA-N
4-6-methyl-2-benzimidazolyl benzamidoxime, afqxausvhhhsmn-uhfffaoysa-n, n-hydroxy-4-6-methyl-1h-benzo d imidazol-2-yl benzimidamide, z-n'-hydroxy-4-5-methyl-1h-benzo d imidazol-2-yl benzimidamide, z-n'-hydroxy-4-5-methyl-3h-1,3-benzodiazol-2-yl benzenecarboximidamide, n'-hydroxy-4-5-methyl-3h-1,3-benzodiazol-2-yl benzenecarboximidamide
73995333
[4-(5-methyl-1,3-dihydrobenzimidazol-2-ylidene)cyclohexa-2,5-dien-1-ylidene]-nitrosomethanamine
CC1=CC2=C(C=C1)NC(=C3C=CC(=C(N)N=O)C=C3)N2
This item is not returnable. View return policy
1256486-22-2
C15H14N4O
266.304
MFCD16658868
SQIGGAWUCHCZKN-UHFFFAOYSA-N
4-6-methyl-2-benzimidazolyl benzamidoxime, afqxausvhhhsmn-uhfffaoysa-n, n-hydroxy-4-6-methyl-1h-benzo d imidazol-2-yl benzimidamide, z-n'-hydroxy-4-5-methyl-1h-benzo d imidazol-2-yl benzimidamide, z-n'-hydroxy-4-5-methyl-3h-1,3-benzodiazol-2-yl benzenecarboximidamide, n'-hydroxy-4-5-methyl-3h-1,3-benzodiazol-2-yl benzenecarboximidamide
73995333
[4-(5-methyl-1,3-dihydrobenzimidazol-2-ylidene)cyclohexa-2,5-dien-1-ylidene]-nitrosomethanamine
CC1=CC2=C(C=C1)NC(=C3C=CC(=C(N)N=O)C=C3)N2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 266.3
Percent Purity 97%
Chemical Name or Material 4-(6-Methyl-2-benzimidazolyl)benzamidoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'