missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diisopropyl-1,1,3,3-tetramethyl-5-indanol, 97%, Thermo Scientific™

Catalog Number p-7025430
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
93892-40-1
C19H30O
274.448
MFCD00061045
WGQDDNQFNLMBOS-UHFFFAOYSA-N
4,6-diisopropyl-1,1,3,3-tetramethyl-5-indanol, 4,6-bis isopropyl-1,1,3,3-tetramethylindan-5-ol, 4,6-diisopropyl-1,1,3,3-tetramethyl-2h-inden-5-ol, 4,6-bis methylethyl-1,1,3,3-tetramethylindan-5-ol, 1,1,3,3-tetramethyl-4,6-di propan-2-yl-2h-inden-5-ol, 1h-inden-5-ol, 2,3-dihydro-1,1,3,3-tetramethyl-4,6-bis 1-methylethyl, 2,3-dihydro-1,1,3,3-tetramethyl-4,6-bis 1-methylethyl-1h-inden-5-ol
621162
1,1,3,3-tetramethyl-4,6-di(propan-2-yl)-2H-inden-5-ol
CC(C)C1=C(C(=C2C(=C1)C(CC2(C)C)(C)C)C(C)C)O
This item is not returnable. View return policy
93892-40-1
C19H30O
274.448
MFCD00061045
WGQDDNQFNLMBOS-UHFFFAOYSA-N
4,6-diisopropyl-1,1,3,3-tetramethyl-5-indanol, 4,6-bis isopropyl-1,1,3,3-tetramethylindan-5-ol, 4,6-diisopropyl-1,1,3,3-tetramethyl-2h-inden-5-ol, 4,6-bis methylethyl-1,1,3,3-tetramethylindan-5-ol, 1,1,3,3-tetramethyl-4,6-di propan-2-yl-2h-inden-5-ol, 1h-inden-5-ol, 2,3-dihydro-1,1,3,3-tetramethyl-4,6-bis 1-methylethyl, 2,3-dihydro-1,1,3,3-tetramethyl-4,6-bis 1-methylethyl-1h-inden-5-ol
621162
1,1,3,3-tetramethyl-4,6-di(propan-2-yl)-2H-inden-5-ol
CC(C)C1=C(C(=C2C(=C1)C(CC2(C)C)(C)C)C(C)C)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 146°C to 148°C
Quantity 1 g
Formula Weight 274.45
Percent Purity 97%
Chemical Name or Material 4,6-Diisopropyl-1,1,3,3-tetramethyl-5-indanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'