Learn More
4,6-Dihydroxypyrimidine, 98%, ACROS Organics™
Brand: Acros Organics 115081000
Code : 29
Additional Details : CAS Number : 1193-24-4 Weight : 0.15000kg
Quantity | 100g |
---|---|
Packaging | Plastic bottle |
Chemical Identifiers
4, 6-Dihydroxypyrimidine | |
97.5 | |
≥97.5% | |
C4H4N2O2 | |
MFCD00016733 |
1193-24-4 | |
100.0 | |
98% | |
112.09 | |
4, 6-Pyrimidinediol |
Specifications
4, 6-Dihydroxypyrimidine | |
100g | |
97.5 | |
Authentic | |
Powder | |
98% | |
112.09 | |
4, 6-Pyrimidinediol |
Yellow to orange | |
1193-24-4 | |
100.0 | |
Plastic bottle | |
≥97.5% | |
C4H4N2O2 | |
MFCD00016733 |